2-Bromo-5-(trifluoromethyl)isonicotinic acid 97%
Lieferant: Apollo Scientific
PC421159-250MG
APOSPC421159-250MGEA
114
EUR
APOSPC421159-250MG
2-Bromo-5-(trifluoromethyl)isonicotinic acid 97%
2-Bromo-5-(trifluoromethyl)isonicotinic acid
Formel:
C₇H₃BrF₃NO₂ Molecular Weight: 270,003 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16610250 CAS-Nummer: 823222-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...