1-Bromo-2-fluoro-3-iodobenzene 98%

Lieferant: Apollo Scientific

PC421173-1G
APOSPC421173-1GEA 0 EUR
APOSPC421173-1G
1-Bromo-2-fluoro-3-iodobenzene 98%
1-Bromo-2-fluoro-3-iodobenzene
Formel: C₆H₃BrFI
Lagertemperatur: Raumtemperatur
MDL: MFCD13194483
CAS-Nummer: 958458-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR