(R)-1-(4-Chloro-3-fluorophenyl)ethane-1,2-diol 95%

Lieferant: Apollo Scientific

PC421192-500MG
APOSPC421192-500MGEA 1030 EUR
APOSPC421192-500MG
(R)-1-(4-Chloro-3-fluorophenyl)ethane-1,2-diol 95%
(R)-1-(4-Chloro-3-fluorophenyl)ethane-1,2-diol
Formel: C₈H₈ClFO₂
Molecular Weight: 190,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29077495
CAS-Nummer: 1453854-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR