2-(Trifluoromethyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC421202-250MG
APOSPC421202-250MGEA
906
EUR
APOSPC421202-250MG
2-(Trifluoromethyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine 95+%
2-(Trifluoromethyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine
Formel:
C₇H₆F₃N₃ Molecular Weight: 189,138 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09954780 CAS-Nummer: 905274-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...