(+/-)-2-(Trifluoromethyl)piperazine 95%

Lieferant: Apollo Scientific

PC421203-100MG PC421203-250MG
APOSPC421203-100MGEA 547 EUR
APOSPC421203-100MG APOSPC421203-250MG
(+/-)-2-(Trifluoromethyl)piperazine 95%
(+/-)-2-(Trifluoromethyl)piperazine
Formel: C₅H₉F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07373382
CAS-Nummer: 131922-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR