2-Bromo-5-fluoro-4-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC421205-1G
APOSPC421205-1GEA 418 EUR
APOSPC421205-1G
2-Bromo-5-fluoro-4-(trifluoromethyl)pyridine 95%
2-Bromo-5-fluoro-4-(trifluoromethyl)pyridine
Formel: C₆H₂BrF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD18259807
CAS-Nummer: 1156542-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR