4-Amino-3-brom-5-fluorbenzonitril 95%

Lieferant: Apollo Scientific

PC421212-5G
APOSPC421212-5GEA 1120 EUR
APOSPC421212-5G
4-Amino-3-brom-5-fluorbenzonitril 95%
4-Amino-3-brom-5-fluorbenzonitril
Formel: C₇H₄BrFN₂
Molecular Weight: 215,0225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059375
CAS-Nummer: 874880-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR