4-Chloro-2-fluoro-N-methylaniline 95%

Lieferant: Apollo Scientific

PC421216-5G
APOSPC421216-5GEA 2900 EUR
APOSPC421216-5G
4-Chloro-2-fluoro-N-methylaniline 95%
4-Chloro-2-fluoro-N-methylaniline
Formel: C₇H₇ClFN
Molecular Weight: 159,589 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11655731
CAS-Nummer: 894099-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR