4-Fluoroquinoline 95%

Lieferant: Apollo Scientific

PC421219-1G
APOSPC421219-1GEA 844 EUR
APOSPC421219-1G
4-Fluoroquinoline 95%
4-Fluoroquinoline
Formel: C₉H₆FN
Lagertemperatur: Raumtemperatur
MDL: MFCD01685510
CAS-Nummer: 394-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR