5,7-Difluoro-4-hydroxyquinoline-3-carbonitrile ≥98%

Lieferant: Apollo Scientific

PC421220-100MG
APOSPC421220-100MGEA 120 EUR
APOSPC421220-100MG
5,7-Difluoro-4-hydroxyquinoline-3-carbonitrile ≥98%
5,7-Difluoro-4-hydroxyquinoline-3-carbonitrile
Formel: C₁₀H₄F₂N₂O
Molecular Weight: 206,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13181113
CAS-Nummer: 541505-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR