6-Cyclopropyl-8-fluoroisoquinolin-1(2H)-one ≥98%

Lieferant: Apollo Scientific

PC421224-50MG
APOSPC421224-50MGEA 446 EUR
APOSPC421224-50MG
6-Cyclopropyl-8-fluoroisoquinolin-1(2H)-one ≥98%
6-Cyclopropyl-8-fluoroisoquinolin-1(2H)-one
Formel: C₁₂H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD23134704
CAS-Nummer: 1242156-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR