6-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 98%

Lieferant: Apollo Scientific

PC421226-100MG PC421226-250MG
APOSPC421226-100MGEA 165 EUR
APOSPC421226-100MG APOSPC421226-250MG
6-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 98%
6-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one
Formel: C₁₀H₁₀FNO
Molecular Weight: 179.191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977761
CAS-Nummer: 145485-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR