Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate 95%

Lieferant: Apollo Scientific
Achtung

PC421234-1G
APOSPC421234-1GEA 59.6 EUR
APOSPC421234-1G
Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate 95%
Methyl 3,3,3-trifluoro-2,2-dimethylpropanoate
Formel: C₆H₉F₃O₂
Molecular Weight: 170,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18253569
CAS-Nummer: 1188911-72-9
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR