3'-Chlor-5'-fluoracetophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4214-5G
APOSPC4214-5GEA
0
EUR
APOSPC4214-5G
3'-Chlor-5'-fluoracetophenon 97%
3'-Chlor-5'-fluoracetophenon
Formel:
C₈H₆ClFO Schmelzpunkt: 51…54 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115834 CAS-Nummer: 842140-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...