4'-Chlor-2'-fluoracetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4215-1G
PC4215-5G
APOSPC4215-1GEA
55.1
EUR
APOSPC4215-1G
APOSPC4215-5G
4'-Chlor-2'-fluoracetophenon 98%
4'-Chlor-2'-fluoracetophenon
Formel:
C₈H₆ClFO Flammpunkt: 95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116328 CAS-Nummer: 175711-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...