Chloro(fluoro)acetyl chloride 96%
Lieferant: Apollo Scientific
|
Gefahr
|
PC4216-1G
APOSPC4216-1GEA
285
EUR
APOSPC4216-1G
Chloro(fluoro)acetyl chloride 96%
Chloro(fluoro)acetyl chloride
Formel:
C₂HCl₂FO Siedepunkt: 69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861134 CAS-Nummer: 359-32-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...