3-Chlor-4-fluorbenzhydrol 97%
Lieferant: Apollo Scientific
PC4224-1G
APOSPC4224-1GEA
429
EUR
APOSPC4224-1G
3-Chlor-4-fluorbenzhydrol 97%
3-Chlor-4-fluorbenzhydrol
Formel:
C₁₃H₁₀ClFO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06201249 CAS-Nummer: 842140-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...