4-(Trifluormethoxy)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4241-25G
PC4241-100G
PC4241-5G
APOSPC4241-25GEA
0
EUR
APOSPC4241-25G
APOSPC4241-100G
APOSPC4241-5G
4-(Trifluormethoxy)phenylboronsäure 98%
4-(Trifluormethoxy)phenylboronsäure
Formel:
C₇H₆BF₃O₃ Schmelzpunkt: 120…127 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01074648 CAS-Nummer: 139301-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...