1-Brom-2,6-difluor-4-nitrobenzol 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1,3-difluoro-5-nitrobenzene

PC4245-1G PC4245-25G PC4245-5G
APOSPC4245-1GEA 48 EUR
APOSPC4245-1G APOSPC4245-25G APOSPC4245-5G
1-Brom-2,6-difluor-4-nitrobenzol 97+%
1-Bromo-2,6-difluoro-4-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Molecular Weight: 237.9864 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04974121
CAS-Nummer: 886762-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR