2-Methoxy-4-(trifluoromethoxy)benzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

PC4266-250MG
APOSPC4266-250MGEA 0 EUR
APOSPC4266-250MG
2-Methoxy-4-(trifluoromethoxy)benzeneboronic acid 95%
2-Methoxy-4-(trifluoromethoxy)benzeneboronic acid
Formel: C₈H₈BF₃O₄
Lagertemperatur: Kühlschrank
MDL: MFCD04037221
CAS-Nummer: 355836-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR