2,4-Difluoro-6-iodoaniline

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Amino-3,5-difluoroiodobenzene

PC4267-1G PC4267-250MG
APOSPC4267-1GEA 37.7 EUR
APOSPC4267-1G APOSPC4267-250MG
2,4-Difluoro-6-iodoaniline
2,4-Difluoro-6-iodoaniline
Formel: C₆H₄F₂IN
Lagertemperatur: Raumtemperatur
MDL: MFCD08059536
CAS-Nummer: 582319-15-1
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR