1-(Trifluoromethylcyclopropyl)benzene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-(Trifluoromethyl)cyclopropyl)benzene

PC4274-250MG
APOSPC4274-250MGEA 34.8 EUR
APOSPC4274-250MG
1-(Trifluoromethylcyclopropyl)benzene 95%
1-(Trifluoromethylcyclopropyl)benzene
Formel: C₁₀H₉F₃
Molecular Weight: 186,18 g/mol
Siedepunkt: 106…107 °C/25mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD08059543
CAS-Nummer: 883547-73-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR