2-(2-Fluorophenyl)-3H-imidazo[4,5-c]pyridine methanesulphonate
Lieferant: Apollo Scientific
PC430102-1G
APOSPC430102-1GEA
568
EUR
APOSPC430102-1G
2-(2-Fluorophenyl)-3H-imidazo[4,5-c]pyridine methanesulphonate
2-(2-Fluorophenyl)-3H-imidazo[4,5-c]pyridine methanesulphonate
Formel:
C₁₂H₈FN₃·CH₃SO₃H Molecular Weight: 309,316 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16040217 CAS-Nummer: 1221449-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...