4,4,4-Trifluoro-1-(3-methoxyphenyl)butane-1,3-dione

Lieferant: Apollo Scientific

PC430107-1G
APOSPC430107-1GEA 79.9 EUR
APOSPC430107-1G
4,4,4-Trifluoro-1-(3-methoxyphenyl)butane-1,3-dione
4,4,4-Trifluoro-1-(3-methoxyphenyl)butane-1,3-dione
Formel: C₁₁H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03419769
CAS-Nummer: 57965-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR