2,3-Difluoro-5-nitrophenol 96%

Lieferant: Apollo Scientific
Achtung

PC430108-250MG PC430108-1G
APOSPC430108-250MGEA 156 EUR
APOSPC430108-250MG APOSPC430108-1G
2,3-Difluoro-5-nitrophenol 96%
2,3-Difluoro-5-nitrophenol
Formel: C₆H₃F₂NO₃
Molecular Weight: 175.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026384
CAS-Nummer: 1119455-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR