Ethyl-5,5,5-trifluoro-4-oxopentanoate

Lieferant: Apollo Scientific

Synonyme: Ethyl 5,5,5-trifluoro-4-oxopentanoate

PC430110-1G
APOSPC430110-1GEA 764 EUR
APOSPC430110-1G
Ethyl-5,5,5-trifluoro-4-oxopentanoate
Ethyl-5,5,5-trifluoro-4-oxopentanoate
Formel: C₇H₉F₃O₃
Molecular Weight: 198,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828184
CAS-Nummer: 70961-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR