Ethyl 3-(benzyl(3-ethoxy-3-oxopropyl)amino)-2,2-difluoropropanoate

Lieferant: Apollo Scientific

PC430111-1G
APOSPC430111-1GEA 466 EUR
APOSPC430111-1G
Ethyl 3-(benzyl(3-ethoxy-3-oxopropyl)amino)-2,2-difluoropropanoate
Ethyl 3-(benzyl(3-ethoxy-3-oxopropyl)amino)-2,2-difluoropropanoate
Formel: C₁₇H₂₃F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD18433564
CAS-Nummer: 851314-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR