2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione
Lieferant: Apollo Scientific
PC430133-1G
APOSPC430133-1GEA
865
EUR
APOSPC430133-1G
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione
Formel:
C₁₁H₈F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19686683 CAS-Nummer: 329710-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...