4-(Trifluoromethyl)piperidin-4-ol hydrochloride

Lieferant: Apollo Scientific

PC430139-1G
APOSPC430139-1GEA 238 EUR
APOSPC430139-1G
4-(Trifluoromethyl)piperidin-4-ol hydrochloride
4-(Trifluoromethyl)piperidin-4-ol hydrochloride
Formel: C₆H₁₀F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD12755740
CAS-Nummer: 1193389-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR