Methyl 3-fluoroisonicotinate

Lieferant: Apollo Scientific

PC430141-1G
APOSPC430141-1GEA 568 EUR
APOSPC430141-1G
Methyl 3-fluoroisonicotinate
Methyl 3-fluoroisonicotinate
Formel: C₇H₆FNO₂
Molecular Weight: 155,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11045048
CAS-Nummer: 876919-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR