1-(tert-Butoxycarbonyl)-4-(4-fluorobenzyl)piperidine-4-carboxylic acid

Lieferant: Apollo Scientific

PC430142-1G
APOSPC430142-1GEA 406 EUR
APOSPC430142-1G
1-(tert-Butoxycarbonyl)-4-(4-fluorobenzyl)piperidine-4-carboxylic acid
1-(tert-Butoxycarbonyl)-4-(4-fluorobenzyl)piperidine-4-carboxylic acid
Formel: C₁₈H₂₄FNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08461311
CAS-Nummer: 906329-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR