tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl (3,3-difluoropiperidin-4-yl)carbamate
PC430144-250MG
APOSPC430144-250MGEA
151
EUR
APOSPC430144-250MG
tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate
tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate
Formel:
C₁₀H₁₈F₂N₂O₂ Molecular Weight: 236,259 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18791569 CAS-Nummer: 1263180-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...