rel-(3R,4R)-4-Benzylamino-1-[tert-butoxycarbonyl]-3-fluoro piperidine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3,4-trans-4-(benzylamino)-3-fluoropiperidine-1-carboxylate racemate

PC430148-1G
APOSPC430148-1GEA 865 EUR
APOSPC430148-1G
rel-(3R,4R)-4-Benzylamino-1-[tert-butoxycarbonyl]-3-fluoro piperidine
rel-(3R,4R)-4-Benzylamino-1-[tert-butoxycarbonyl]-3-fluoro piperidine
Formel: C₁₇H₂₅FN₂O₂
Molecular Weight: 308.4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20261376
CAS-Nummer: 211108-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR