tert-Butyl 3,4-cis-4-amino-3-fluoropiperidine-1-carboxylate racemate
Lieferant: Apollo Scientific
|
Gefahr
|
PC430151-1G
APOSPC430151-1GEA
575
EUR
APOSPC430151-1G
tert-Butyl 3,4-cis-4-amino-3-fluoropiperidine-1-carboxylate racemate
tert-Butyl 3,4-cis-4-amino-3-fluoropiperidine-1-carboxylate racemate
Formel:
C₁₀H₁₉FN₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD12912699 CAS-Nummer: 577691-56-6 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...