cis-1-Boc-3-fluoro-4-hydroxypiperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: (3.4)-cis-3-Fluoro-4-hydroxy-piperidine-1-carboxylic acid tert-Butyl ester, cis-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

PC430153-1G
APOSPC430153-1GEA 36.4 EUR
APOSPC430153-1G
cis-1-Boc-3-fluoro-4-hydroxypiperidine
cis-1-Boc-3-fluoro-4-hydroxypiperidine
Formel: C₁₀H₁₈FNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD18791209
CAS-Nummer: 955028-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR