Ethyl 1-benzyl-5,5-difluoro-4-hydroxypiperidine-3-carboxylate

Lieferant: Apollo Scientific

PC430163-1G
APOSPC430163-1GEA 655 EUR
APOSPC430163-1G
Ethyl 1-benzyl-5,5-difluoro-4-hydroxypiperidine-3-carboxylate
Ethyl 1-benzyl-5,5-difluoro-4-hydroxypiperidine-3-carboxylate
Formel: C₁₅H₁₉F₂NO₃
Molecular Weight: 299,313 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21601887
CAS-Nummer: 1356339-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR