1-tert-Butyl-3-ethyl-5,5-difluoro-4-oxopiperidine-1,3-dicarboxylate

Lieferant: Apollo Scientific

Synonyme: 1-tert-Butyl 3-ethyl 5,5-difluoro-4-oxopiperidine-1,3-dicarboxylate

PC430165-1G
APOSPC430165-1GEA 655 EUR
APOSPC430165-1G
1-tert-Butyl-3-ethyl-5,5-difluoro-4-oxopiperidine-1,3-dicarboxylate
1-tert-Butyl-3-ethyl-5,5-difluoro-4-oxopiperidine-1,3-dicarboxylate
Formel: C₁₃H₁₉F₂NO₅
Molecular Weight: 307,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21601893
CAS-Nummer: 1356338-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR