2,3-Difluoro-4-nitrophenol

Lieferant: Apollo Scientific

PC430167-1G
APOSPC430167-1GEA 568 EUR
APOSPC430167-1G
2,3-Difluoro-4-nitrophenol
2,3-Difluoro-4-nitrophenol
Formel: C₆H₃F₂NO₃
Molecular Weight: 175,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185572
CAS-Nummer: 123173-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR