tert-Butyl-trans-3-(tert-butoxycarbonyloxy)-4-fluoropyrrolidine-1-carboxylate
Lieferant: Apollo Scientific
PC430191-1G
APOSPC430191-1GEA
865
EUR
APOSPC430191-1G
tert-Butyl-trans-3-(tert-butoxycarbonyloxy)-4-fluoropyrrolidine-1-carboxylate
tert-Butyl-trans-3-(tert-butoxycarbonyloxy)-4-fluoropyrrolidine-1-carboxylate
Formel:
C₁₄H₂₄FNO₅ Molecular Weight: 305,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22199429 CAS-Nummer: 1373503-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...