N-[2-Nitro-4-(trifluormethyl)phenyl]piperidin 97%

Lieferant: Apollo Scientific
Achtung

PC4302-1G PC4302-5G
APOSPC4302-1GEA 0 EUR
APOSPC4302-1G APOSPC4302-5G
N-[2-Nitro-4-(trifluormethyl)phenyl]piperidin 97%
N-[2-Nitro-4-(trifluormethyl)phenyl]piperidin
Formel: C₁₂H₁₃F₃N₂O₂
Schmelzpunkt: 48…50 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052908
CAS-Nummer: 1692-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR