8-Boc-2-fluoro-8-azabicyclo[3.2.1]octan-3-ol

Lieferant: Apollo Scientific

PC430202-1G
APOSPC430202-1GEA 865 EUR
APOSPC430202-1G
8-Boc-2-fluoro-8-azabicyclo[3.2.1]octan-3-ol
8-Boc-2-fluoro-8-azabicyclo[3.2.1]octan-3-ol
Formel: C₁₂H₂₀FNO₃
Molecular Weight: 245,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22580393
CAS-Nummer: 1404196-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR