3-Acetamido-2-nitrophenyl 4-(benzyl(methyl)amino)-3,5-difluorobenzoate

Lieferant: Apollo Scientific

PC430219-1G
APOSPC430219-1GEA 655 EUR
APOSPC430219-1G
3-Acetamido-2-nitrophenyl 4-(benzyl(methyl)amino)-3,5-difluorobenzoate
3-Acetamido-2-nitrophenyl 4-(benzyl(methyl)amino)-3,5-difluorobenzoate
Formel: C₂₃H₁₉F₂N₃O₅
Molecular Weight: 455.411 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27978602
CAS-Nummer: 1864061-72-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR