tert-Butyl 3,3-difluoro-4-hydroxypyrrolidine-1-carboxylate
Lieferant: Apollo Scientific
PC430237-250MG
PC430237-1G
APOSPC430237-250MGEA
285
EUR
APOSPC430237-250MG
APOSPC430237-1G
tert-Butyl 3,3-difluoro-4-hydroxypyrrolidine-1-carboxylate
tert-Butyl 3,3-difluoro-4-hydroxypyrrolidine-1-carboxylate
Formel:
C₉H₁₅F₂NO₃ Molecular Weight: 223.217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27956892 CAS-Nummer: 1434141-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...