Ethyl 2,2-difluoro-3-hydroxy-3-phenylpropanoate

Lieferant: Apollo Scientific

PC430249-1G
APOSPC430249-1GEA 326 EUR
APOSPC430249-1G
Ethyl 2,2-difluoro-3-hydroxy-3-phenylpropanoate
Ethyl 2,2-difluoro-3-hydroxy-3-phenylpropanoate
Formel: C₁₁H₁₂F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00725033
CAS-Nummer: 92207-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR