Ethyl 2,2-difluoro-3-(4-fluorophenyl)-3-hydroxypropanoate
Lieferant: Apollo Scientific
PC430250-1G
APOSPC430250-1GEA
466
EUR
APOSPC430250-1G
Ethyl 2,2-difluoro-3-(4-fluorophenyl)-3-hydroxypropanoate
Ethyl 2,2-difluoro-3-(4-fluorophenyl)-3-hydroxypropanoate
Formel:
C₁₁H₁₁F₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12403972 CAS-Nummer: 321856-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...