9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide
Lieferant: Apollo Scientific
PC430251-1G
APOSPC430251-1GEA
79.9
EUR
APOSPC430251-1G
9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide
9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide
Formel:
C₂₀H₁₉BrF₃NO Molecular Weight: 426,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27978674 CAS-Nummer: 182438-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...