tert-Butyl 4-(4'-(trifluoromethyl)biphenyl-2-ylcarboxamido)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430252-1G
APOSPC430252-1GEA 655 EUR
APOSPC430252-1G
tert-Butyl 4-(4'-(trifluoromethyl)biphenyl-2-ylcarboxamido)piperidine-1-carboxylate
tert-Butyl 4-(4'-(trifluoromethyl)biphenyl-2-ylcarboxamido)piperidine-1-carboxylate
Formel: C₂₄H₂₇F₃N₂O₃
Molecular Weight: 448,478 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15983728
CAS-Nummer: 1389198-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR