N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide hydrochloride

Lieferant: Apollo Scientific

PC430253-1G
APOSPC430253-1GEA 865 EUR
APOSPC430253-1G
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide hydrochloride
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide hydrochloride
Formel: C₁₉H₁₉F₃N₂O·ClH
Molecular Weight: 384,823 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28016245
CAS-Nummer: 1838769-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR