N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide
Lieferant: Apollo Scientific
PC430254-1G
APOSPC430254-1GEA
865
EUR
APOSPC430254-1G
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide
Formel:
C₁₉H₁₉F₃N₂O Molecular Weight: 348,362 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27978680 CAS-Nummer: 182439-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...