1-tert-Butyl 3-Methyl 5,5-difluoro-4-hydroxypiperidine-1,3-dicarboxylate

Lieferant: Apollo Scientific

PC430260-1G
APOSPC430260-1GEA 865 EUR
APOSPC430260-1G
1-tert-Butyl 3-Methyl 5,5-difluoro-4-hydroxypiperidine-1,3-dicarboxylate
1-tert-Butyl 3-Methyl 5,5-difluoro-4-hydroxypiperidine-1,3-dicarboxylate
Formel: C₁₂H₁₉F₂NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD28155093
CAS-Nummer: 1864014-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR